missing translation for 'onlineSavingsMsg'
Learn More

Barium 1-methoxy-2-propoxide, 25% w/w in 1-methoxy-2-propanol, Thermo Scientific™

Product Code. 11379627
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379627 5 g 5g
11369627 25 g 25g
11359627 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11379627 Supplier Thermo Scientific Alfa Aesar Supplier No. 041257.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130812-47-4
Molecular Formula C8H18BaO4
Molecular Weight (g/mol) 315.555
MDL Number MFCD00078021
InChI Key WCXRGQPXPCQKSJ-UHFFFAOYSA-N
Synonym bis 2-ethoxyethoxy barium, barium methoxypropoxide w/w in methoxypropanol, barium bis 2-ethoxyethan-1-olate, barium 2+ ion bis 2-ethoxyethanolate
PubChem CID 14619883
IUPAC Name barium(2+);2-ethoxyethanolate
SMILES CCOCC[O-].CCOCC[O-].[Ba+2]

Specifications

Density 1.04 g/mL
Flash Point 35°C (95°F)
Linear Formula Ba[OCH(CH3)CH2OCH3]2
Quantity 5 g
UN Number UN2924
Sensitivity Air and moisture sensitive
Formula Weight 315.55
Concentration or Composition (by Analyte or Components) 25% w/w in 1-methoxy-2-propanol
Physical Form Liquid
Chemical Name or Material Barium 1-methoxy-2-propoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.