missing translation for 'onlineSavingsMsg'
Learn More

Baricitinib phosphate, MedChemExpress

Product Code. 16476448
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476448 10 mg 10mg
16449037 5 mg 5mg
16412557 50 mg 50mg
16436736 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476448 Supplier Medchem Express Supplier No. HY15315A
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Baricitinib phosphate (LY3009104 phosphate; INCB028050 phosphate) is a selective orally bioavailable JAK1/JAK2 inhibitor with IC50 of 5.9 nM and 5.7 nM, respectively.

Baricitinib phosphate (LY3009104 phosphate

Specifications

Chemical Name or Material Baricitinib phosphate
CAS 1187595-84-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H20N7O6PS
Quantity 10 mg
Synonym LY3009104 phosphateINCB028050 phosphate
Solubility Information DMSO : ≥ 4.7 mg/mL (10.01 mM)
SMILES N#CCC1(N2N=CC(C3=C4C(NC=C4)=NC=N3)=C2)CN(S(=O)(CC)=O)C1.O=P(O)(O)O
Molecular Weight (g/mol) 469.41
Formula Weight 469.41
Percent Purity 99.29%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.