missing translation for 'onlineSavingsMsg'
Learn More

Baricitinib, MedChemExpress

Product Code. 16466448
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16466448 10 mg 10mg
16439037 5 mg 5mg
16426736 100 mg 100mg
16402557 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16466448 Supplier Medchem Express Supplier No. HY15315
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Baricitinib (LY3009104; INCB028050) is a selective and orally bioavailable JAK1 and JAK2 inhibitor with IC50s of 5.9 nM and 5.7 nM, respectively.

Baricitinib (LY3009104

Specifications

Chemical Name or Material Baricitinib
CAS 1187594-09-7
Color White
Purity Grade Notes Research
Molecular Formula C16H17N7O2S
Quantity 10 mg
Synonym LY3009104 INCB028050
Solubility Information DMSO : 25 mg/mL (67.31 mM; Need ultrasonic and warming)
SMILES N#CCC1(N2N=CC(C3=C4C(NC=C4)=NC=N3)=C2)CN(S(=O)(CC)=O)C1
Molecular Weight (g/mol) 371.42
Formula Weight 371.42
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.