Learn More
Masterflex™ BarbLock Assemblies
Product Code. 11759729
Change view
Click to view available options
For Use With:
Fits 1/4 in. and L/S 15, 17, & 24 Tubing
Fits 1/4 in. x 3/8 in. Tubing
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
11759729 Fits 1/4 in. x 3/8 in. Tubing Each
15360484 Fits 1/4 in. and L/S 15, 17, & 24 Tubing Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11759729 Supplier Masterflex™ Supplier No. 3060400
Only null left
Add to basket
Add to basket
Provide uniform 360-degree radial crimping for a leak-free, durable seal. Masterflex™ Barblock Assemblies feature an easy to install, three-piece locking mechanism ensures the tubing and hose barb won't disengage.
  • Safe to use with pressures up to 140psi or upper pressure limit of tubing
  • An excellent alternative to securing tubing and hose barbs with cable ties — no more crimps or tube distortion
  • BarbLock assembly consists of three pieces: the sleeve, collect, and barbed fittings
  • To install the fitting, slide the sleeve and collet onto the tubing and then inserts the barbed fitting into the tubing
  • Prelock the sleeve and collet by pressing them together until you hear a click
  • Slide the sleeve and collet to the edge of the barbed fitting and lock with the BarbLock tool
  • Once assembled, the BarbLock assembly can only be unlocked with the removal tool
Specifications
For Use With Fits 1/4 in. x 3/8 in. Tubing
For Use With (Application) Easy to install, three-piece locking mechanism ensures the tubing and hose barb won't disengage
Packaging Quantity 1 Pack
Product Type BarbLock Assembly