missing translation for 'onlineSavingsMsg'
Learn More

Barbituric acid, 99%

Product Code. 11444300
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444300 100 g 100g
11434300 25 g 25g
11454300 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444300 Supplier Thermo Scientific Alfa Aesar Supplier No. A10934.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67-52-7 | C4H4N2O3 | 128.09 g/mol

Barbituric acid is widely used in the manufacturing of plastics, textiles, polymers and pharmaceuticals. It is an active ingredient in the production of Vitamin B2. It is a strong acid in an aqueous medium with an active methylene group involved Knoevenegal condensation. It is used as precursor for the preparation of 5-arylidene barbituric acid by reacting with aromatic aldehyde. It is also used in electrochemical oxidation of iodine using cyclic voltammetry and controlled potential coulometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Barbituric acid is widely used in the manufacturing of plastics, textiles, polymers and pharmaceuticals. It is an active ingredient in the production of Vitamin B2. It is a strong acid in an aqueous medium with an active methylene group involved Knoevenegal condensation. It is used as precursor for the preparation of 5-arylidene barbituric acid by reacting with aromatic aldehyde. It is also used in electrochemical oxidation of iodine using cyclic voltammetry and controlled potential coulometry.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-52-7
Molecular Formula C4H4N2O3
Molecular Weight (g/mol) 128.09
MDL Number MFCD00006666
InChI Key HNYOPLTXPVRDBG-UHFFFAOYSA-N
Synonym barbituric acid, malonylurea, 2,4,6 1h,3h,5h-pyrimidinetrione, 6-hydroxyuracil, pyrimidinetrione, 2,4,6-trihydroxypyrimidine, 2,4,6-pyrimidinetrione, pyrimidine-2,4,6 1h,3h,5h-trione, barbiturate, malonylharnstoff
PubChem CID 6211
ChEBI CHEBI:16294
IUPAC Name 1,3-diazinane-2,4,6-trione
SMILES O=C1CC(=O)NC(=O)N1

Specifications

Melting Point ∼250°C (decomposition)
pH 2 to 3
Flash Point 150°C (302°F)
Quantity 100 g
Beilstein 120502
Merck Index 14,963
Solubility Information Slightly soluble in water.
Formula Weight 128.09
Percent Purity 99%
Chemical Name or Material Barbituric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.