Learn More
Masterflex™ Barbed NPT Male Pipe Adapter Elbows
Product Code. 15385696 Shop All Masterflex Products
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Pack of 5
Product Code. Material Unit Size
15385696 Polypropylene Pack of 5
1 options
Product Code. 15385696 Supplier Masterflex™ Supplier No. 4061069
  • ONLINE EXCLUSIVE PRICE
    £19.00
    £18.03 / Pack of 5
    Save £0.97
    5% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ideal for low-pressure applications
  • These economical barbed fittings easily connect fluid paths without the use of tools.
  • Just slip your flexible tubing or hose onto the barb to create a secure seal.
  • Ideal for low-pressure applications.
  • Chemically resistant.
  • Available in a variety of sizes, materials, and configurations.
Specifications
Description Barb x NPT(M)
Material Polypropylene
Sterilization Method Ethylene Oxide, Gamma
Product Type Barbed NPT male pipe adapter elbows, Natural PP, 3/4 in. NPT x 3/4 in.