missing translation for 'onlineSavingsMsg'
Learn More

Barasertib, MedChemExpress

Product Code. 16489846
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16489846

Brand: Medchem Express HY10127

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Barasertib (AZD1152), a pro-drug of Barasertib-hQPA, is a highly selective Aurora B inhibitor with an IC50 of 0.37 nM in a cell-free assay. Barasertib (AZD1152) induces growth arrest and apoptosis in cancer cells.

Barasertib (AZD1152), a pro-drug of Barasertib-hQPA, is a highly selective Aurora B inhibitor with an IC50 of 0.37 nM in a cell-free assay. Barasertib (AZD1152) induces growth arrest and apoptosis in cancer cells.

Specifications

Chemical Name or Material Barasertib
CAS 722543-31-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H31FN7O6P
Quantity 50 mg
Synonym AZD1152
Solubility Information DMSO : ≥ 33 mg/mL (56.17 mM)
SMILES O=C(NC1=CC=CC(F)=C1)CC2=CC(NC3=C4C=CC(OCCCN(CC)CCOP(O)(O)=O)=CC4=NC=N3)=NN2
Molecular Weight (g/mol) 587.54
Formula Weight 587.54
Percent Purity 95.5%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.