missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAPTA, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic Acid |
| CAS | 85233-19-8 |
| Molecular Formula | C22 H24 N2 O10 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C22H24N2O10/c25-19(26)11-23(12-20(27)28)15-5-1-3-7-17(15)33-9-10-34-18-8-4-2-6-16(18)24(13-21(29)30)14-22(31)32/h1-8H,9-14H2,(H,25,26)(H,27,28)(H,29,30)(H,31,32) |
| SMILES | OC(=O)CN(CC(=O)O)c1ccccc1OCCOc2ccccc2N(CC(=O)O)CC(=O)O |
| IUPAC Name | 2-[2-[2-[2-[bis(carboxymethyl)amino]phenoxy]ethoxy]-N-(carboxymethyl)anilino]acetic acid |
| Molecular Weight (g/mol) | 476.43 |
| Formula Weight | 476.1431 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction