missing translation for 'onlineSavingsMsg'
Learn More

BAPTA, MedChemExpress

Product Code. 16451516
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451516 500 mg 500mg
16424246 100 mg 100mg
16400906 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16451516 Supplier Medchem Express Supplier No. HY100168
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress BAPTA is a non-permeable, selective extracellular calcium chelator, with 105-fold greater affinity for Ca2+ than Mg2+. BAPTA is a valuable tool to study the role of calcium in cell signaling.

BAPTA is a non-permeable, selective extracellular calcium chelator, with 105-fold greater affinity for Ca2+ than Mg2+. BAPTA is a valuable tool to study the role of calcium in cell signaling.

Specifications

Chemical Name or Material BAPTA
CAS 85233-19-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H24N2O10
Quantity 500 mg
Solubility Information 1M NaOH : 100 mg/mL (209.89 mM; ultrasonic and adjust pH to 11 with NaOH) ∣DMSO : 15.62 mg/mL (32.79 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(O)CN(C1=CC=CC=C1OCCOC2=CC=CC=C2N(CC(O)=O)CC(O)=O)CC(O)=O
Molecular Weight (g/mol) 476.43
Formula Weight 476.43
Percent Purity 97.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.