missing translation for 'onlineSavingsMsg'
Learn More

BAPTA-AM, MedChemExpress

Product Code. 16453078
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453078

Brand: Medchem Express HY100545

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BAPTA-AM is a well-known membrane permeable Ca2+ chelator. BAPTA-AM inhibits hERG channels, hKv1.3 and hKv1.5 channels in HEK 293 cells with IC50s of 1.3 μM, 1.45 μM and 1.23 μM, respectively.

BAPTA-AM is a well-known membrane permeable Ca2+ chelator. BAPTA-AM inhibits hERG channels, hKv1.3 and hKv1.5 channels in HEK 293 cells with IC50s of 1.3 μM, 1.45 μM and 1.23 μM, respectively.

Specifications

Chemical Name or Material BAPTA-AM
CAS 126150-97-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C34H40N2O18
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (65.39 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C(OCOC(C)=O)CN(C(C=CC=C1)=C1OCCOC(C=CC=C2)=C2N(CC(OCOC(C)=O)=O)CC(OCOC(C)=O)=O)CC(OCOC(C)=O)=O
Molecular Weight (g/mol) 764.68
Formula Weight 764.68
Percent Purity 98.38%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.