missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BAP31 Recombinant Rabbit Monoclonal Antibody (23GB3680), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | BAP31 |
|---|---|
| Clone | 23GB3680 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088676
![]() |
Invitrogen™
MA553450 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088003
|
Invitrogen™
MA553451 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAP31 Recombinant Monoclonal antibody specifically detects BAP31 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| BAP31 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P51572, Q61335 | |
| 10134, 27061 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3680 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 6C6 antigen; 6C6-AG; 6C6-AG tumor-associated antigen; accessory protein BAP31; B cell receptor associated protein 31; BAP31; Bcap31; B-cell receptor-associated protein 31; BCR-associated protein 31; BCR-associated protein Bap31; CDM; CDM protein; DDCH; DXS1357E; hypothetical protein LOC533949; OTTHUMP00000025978; p28; p28 Bap31; Protein CDM | |
| A synthetic peptide of human DHFR (150-200AA). | |
| Primary | |
| -20°C | |
| BCAP31 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title