missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAP1 Antibody (BAP1/13501R), Novus Biologicals™
Rabbit Monoclonal Antibody
£219.00 - £392.00
Specifications
| Antigen | BAP1 |
|---|---|
| Clone | BAP1/13501R |
| Concentration | 0.2 mg/ml |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30287648
|
Novus Biologicals
NBP3-44625-20ug |
20 μg |
£219.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30303321
|
Novus Biologicals
NBP3-44625-100ug |
100 μg |
£392.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAP1 Monoclonal antibody specifically detects BAP1 in Human samples. It is validated for ImmunohistochemistrySpecifications
| BAP1 | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2 | |
| Recombinant fragment (around aa400-729) of human BAP1 protein (Uniprot: Q92560). Exact sequence is proprietary. | |
| Primary | |
| Store at 4⪚C. |
| BAP1/13501R | |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Tumor Suppressors | |
| 10 mM PBS with 0.05% BSA | |
| 8314 | |
| IgG κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title