missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Baohuoside I

Product Code. 16478818
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478818 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16478818 Supplier MedChemExpress Supplier No. HYN0011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Baohuoside I, a flavonoid isolated from Epimedium koreanum Nakai, acts as an inhibitor of CXCR4, downregulates CXCR4 expression, induces apoptosis and shows anti-tumor activity.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 514.52 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 113558-15-9
Product Type Natural Products
Purity 98.37%
Solubility DMSO : ≥ 32 mg/mL (62.19 mM)
Synonym Icariin-II∣ Icariside-II
Target CXCR∣Apoptosis
Quantity 10 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.