missing translation for 'onlineSavingsMsg'
Learn More

Banoxantrone dihydrochloride, MedChemExpress

Product Code. 16493366
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493366 25 mg 25mg
16425758 10 mg 10mg
16408437 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16493366 Supplier Medchem Express Supplier No. HY13562A
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Banoxantrone dihydrochloride is a novel bioreductive agent that can be reduced to a stable, DNA-affinic compound AQ4, which is a potent topoisomerase II inhibitor.

Banoxantrone dihydrochloride is a novel bioreductive agent that can be reduced to a stable, DNA-affinic compound AQ4, which is a potent topoisomerase II inhibitor.

Specifications

Chemical Name or Material Banoxantrone dihydrochloride
CAS 252979-56-9
Color Dark blue
Purity Grade Notes Research
Molecular Formula C22H30Cl2N4O6
Quantity 25 mg
Synonym AQ4N dihydrochloride
Solubility Information H2O : 25 mg/mL (48.32 mM; Need ultrasonic and warming) ∣DMSO : 12.5 mg/mL (24.16 mM; Need ultrasonic)
SMILES O=C1C2=C(C(O)=CC=C2O)C(C3=C(NCC[N+](C)(C)[O-])C=CC(NCC[N+](C)(C)[O-])=C13)=O.Cl.Cl
Molecular Weight (g/mol) 517.4
Formula Weight 517.4
Percent Purity 98.78%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.