missing translation for 'onlineSavingsMsg'
Learn More

Banoxantrone dihydrochloride, MedChemExpress

Product Code. 16425758
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16425758

Brand: Medchem Express HY13562A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Banoxantrone dihydrochloride is a novel bioreductive agent that can be reduced to a stable, DNA-affinic compound AQ4, which is a potent topoisomerase II inhibitor.

Banoxantrone dihydrochloride is a novel bioreductive agent that can be reduced to a stable, DNA-affinic compound AQ4, which is a potent topoisomerase II inhibitor.

Specifications

Chemical Name or Material Banoxantrone dihydrochloride
CAS 252979-56-9
Color Dark blue
Purity Grade Notes Research
Molecular Formula C22H30Cl2N4O6
Quantity 10 mg
Synonym AQ4N dihydrochloride
Solubility Information H2O : 25 mg/mL (48.32 mM; Need ultrasonic and warming) ∣DMSO : 12.5 mg/mL (24.16 mM; Need ultrasonic)
SMILES O=C1C2=C(C(O)=CC=C2O)C(C3=C(NCC[N+](C)(C)[O-])C=CC(NCC[N+](C)(C)[O-])=C13)=O.Cl.Cl
Molecular Weight (g/mol) 517.4
Formula Weight 517.4
Percent Purity 98.78%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.