missing translation for 'onlineSavingsMsg'
Learn More

Banoxantrone dihydrochloride

Product Code. 18738556
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18738556 25 mg 25mg
18706416 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18738556 Supplier Tocris Bioscience Supplier No. 4219/25

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prodrug topoisomerase II inhibitor

Banoxantrone dihydrochloride is a bioreductive prodrug; activated by hypoxia to yield the topoisomerase II inhibitor AQ4. Displays little to no cytotoxic activity against 60 tumor cell lines tested under oxic conditions (IC50 >100 μM). Exhibits anti-tumor efficacy in vivo when combined with oxic cell cytotoxins; enhances the anti-tumor effect of cyclophosphamide (Cat. No. 4091) in a mouse tumor model.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,4-Bis[[2-(dimethyloxidoamino)ethyl]amino]-5,8-dihydroxy-9,10-anthracenedione dihydrochloride
CAS 252979-56-9
Assay Percent Range >98%
Quantity 25 mg
Product Line Tocris
Molecular Formula C22H28N4O6.2HCl
Formula Weight Observed MW: 517.4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.