missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BANK1 Polyclonal Antibody

Description
Immunogen sequence: RHGHKELKKI FEDFSIQEID INNEQENDYE EDIASFSTYI PSTQNPAFHH ESRKTYGQSA DGAEANEMEG EGKQNGSGME TKHSPL Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 55%.
The protein encoded by this gene is a B-cell-specific scaffold protein that functions in B-cell receptor-induced calcium mobilization from intracellular stores. This protein can also promote Lyn-mediated tyrosine phosphorylation of inositol 1,4,5-trisphosphate receptors. Polymorphisms in this gene are associated with susceptibility to systemic lupus erythematosus. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
| Antigen | BANK1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | BANK1 |
| Gene Accession No. | Q8NDB2 |
| Gene Alias | A530094C12Rik; AI451642; AVIEF; B cell scaffold protein with ankyrin repeats 1; BANK; Bank1; B-cell scaffold protein with ankyrin repeats; B-cell scaffold protein with ankyrin repeats 1; B-cell scaffold protein with ankyrin repeats 1 delta 2 transcript; LRRGT00104; Protein AVIEF |
| Gene Symbols | BANK1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?