missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BANF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570404
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 93%.
BANF2 gene ontology annotations related to this gene include DNA binding.
Specifications
| BANF2 | |
| Polyclonal | |
| Unconjugated | |
| BANF2 | |
| 4930517K23Rik; BAF2; BAFL; BAF-L; Baf-like; BANF family member 2; Banf2; barrier to autointegration factor 2; barrier-to-autointegration factor 2; Barrier-to-autointegration factor-like protein; C20orf179; Gm115; likely ortholog of H. sapiens Chromosome 20 open reading frame 179 (C20orf179); LOC538599 protein; RGD1307909; testis secretory sperm-binding protein Li 212a | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 403171 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8BVR0 | |
| BANF2 | |
| synthetic peptide directed towards the middle region of human BANF2 (aa 2-51). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction