missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BANF1, Mouse anti-Human, Clone: M2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00008815-M07.100ug
This item is not returnable.
View return policy
Description
Sequence: MTTSQKHRDFVAEPMGEKPVGSLAGIGEVLGKKLEERGFDKAYVVLGQFLVLKKDEDLFREWLKDTCGANAKQSRDCFGCLREWCDAFLSpecifications
| BANF1 | |
| Monoclonal | |
| Unconjugated | |
| barrier to autointegration factor 1 | |
| BAF/BCRP1/D14S1460/MGC111161 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| M2 | |
| In 1x PBS, pH 7.4 | |
| BC005942 | |
| BANF1 | |
| BANF1 (AAH05942, 1 a.a. ∼ 89 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 8815 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction