missing translation for 'onlineSavingsMsg'
Learn More

BANF1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18420967 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420967 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420967 Supplier Novus Biologicals Supplier No. NBP176750MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BANF1 Polyclonal antibody specifically detects BANF1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen BANF1
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias BAFMGC111161, barrier to autointegration factor 1, barrier-to-autointegration factor, BCRG1, BCRP1, Breakpoint cluster region protein 1, D14S1460
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 15 amino acids near the carboxy terminus of human BANF1. The immunogen is located within the last 50 amino acids of BANF1. Amino Acid Sequence: LKKDEDLFREWLKDT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 8815
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.