missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BANF1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76750UV
This item is not returnable.
View return policy
Description
BANF1 Polyclonal antibody specifically detects BANF1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| BANF1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 350 | |
| BAFMGC111161, barrier to autointegration factor 1, barrier-to-autointegration factor, BCRG1, BCRP1, Breakpoint cluster region protein 1, D14S1460 | |
| Antibody was raised against a peptide corresponding to 15 amino acids near the carboxy terminus of human BANF1. The immunogen is located within the last 50 amino acids of BANF1. Amino Acid Sequence: LKKDEDLFREWLKDT | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 8815 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction