missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bandmate™ Reagent Tubes

Product Code. 17193309 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 tubes
500 tubes
Unit Size:
Pack of 100
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17193309 500 tubes Pack of 500
17103319 100 tubes Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17193309 Supplier Invitrogen™ Supplier No. BW020X25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bandmate Reagent Tubes are specially designed for use with the Bandmate Automated Western Blot Processor.

The tubes are required for dispensing primary and secondary antibodies into the Bandmate mini and midi incubation trays during the automated western blot processing procedure. Two reagent tubes are required for each blot.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Description Bandmate Reagent Tubes, 500 Pack
For Use With (Application) Western Blot
For Use With (Equipment) Invitrogen Bandmate Automated Western Blot Processor
Product Line Bandmate
Type Reagent Tube
Quantity 500 tubes
Shipping Condition Ambient
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.