missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bandmate™ 4L Nalgene™ Bottle
Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 bottle
Unit Size:
Each
Description
The bottle is used for storing wash buffer used throughout western blot processing. Each package contains one 4L bottle with quick connection tubing.
Specifications
Specifications
| Color | Clear |
| Description | 4L Nalge Bottle |
| For Use With (Application) | Western Blot |
| For Use With (Equipment) | Invitrogen Bandmate Automated Western Blot Processor |
| Product Line | Bandmate |
| Type | Bottle |
| Quantity | 1 bottle |
| Shipping Condition | Ambient |
| Dimensions (L x W x H) | 25.654 x 21.59 x 6.045 cm |
| Weight | 0.227 kg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction