missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Band 3 Recombinant Rabbit Monoclonal Antibody (23GB4625), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552717
This item is not returnable.
View return policy
Description
The Band 3 protein is the most abundant integral protein of human erythrocyte membranes and functions as the major anion transporter polypeptide of erythrocytes. Band 3 is a 90-100 kDa protein, its microheterogeneity is ascribed mainly to the structural heterogeneity of the carbohydrate moiety. The C-terminal portion of the molecule spans the membrane several times, and is responsible for anion transport. The N-terminal portion, comprising almost half of the polypeptide chain of Band 3, is located inside the red cell and interacts with cytoskeletal and cytoplasmic proteins such as ankyrin.
Specifications
| Band 3 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P02730, P04919, P23562 | |
| SLC4A1 | |
| A synthesized peptide derived from human Band 3/CD233 (20-70AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 23GB4625 | |
| Unconjugated | |
| SLC4A1 | |
| AE 1; AE1; anion exchange protein 1; anion exchanger 1; anion exchanger-1; band 3 anion transport protein; BND3; CAMP; CD233; CHC; DI; Diego blood group; EMPB3; EPB3; erythrocyte membrane protein band 3; erythroid anion exchange protein; FR; Froese blood group; l11Jus51; MEB3; MGC116750; MGC116753; MGC126619; MGC126623; RTA1A; SAO; Slc4a1; solute carrier family 4 (anion exchanger), member 1; solute carrier family 4 (anion exchanger), member 1 (Diego blood group); solute carrier family 4 member 1; solute carrier family 4 member 1 (Diego blood group); solute carrier family 4, anion exchanger, member 1 (erythrocyte membrane protein band 3, Diego blood group); solute carrier family 4, anion exchanger, number 1; Solute carrier family 4, member 1, anion exchange protein 1 (kidney band 3); SPH4; SW; Swann blood group; Waldner blood group; WD; WD1; WR; Wright blood group; ZNF828 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20533, 24779, 6521 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction