missing translation for 'onlineSavingsMsg'
Learn More

Band 3 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18322256 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322256 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322256 Supplier Novus Biologicals Supplier No. NBP299575JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Band 3 Polyclonal specifically detects Band 3 in Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Band 3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias AE 1, AE1MGC126619, Anion exchange protein 1, Anion exchanger 1, band 3 anion transport protein, BND3, CD233, CD233 antigen, DI, EMPB3, EPB3MGC126623, erythrocyte membrane protein band 3, erythroid anion exchange protein, FR, Froese blood group, MGC116750, MGC116753, RTA1A, Solute carrier family 4 member 1, solute carrier family 4, anion exchanger, member 1 (erythrocyte membraneprotein band 3, Diego blood group), solute carrier family 4, anion exchanger, number 1, SW, Swann blood group, Waldner blood group, WD, WD1, WR, Wright blood group
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of rat Band 3.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 6521
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.