missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Band 3/AE 1 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
£119.00 - £330.00
Specifications
| Antigen | Band 3/AE 1 |
|---|---|
| Clone | 3K21 |
| Concentration | 800 μg/mL |
| Applications | Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236585
|
Proteintech
81066-1-RR-100UL |
100 μL |
£360.00 £330.00 / 100µL Save £30.00 8% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235740
|
Proteintech
81066-1-RR-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Band 3/AE 1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Band 3/AE 1. This antibody recognizes human antigen. The Band 3/AE 1 antibody has been validated for the following applications: IHC, IF-P, ELISA analysis.Specifications
| Band 3/AE 1 | |
| 800 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 6521 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 3K21 | |
| Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| AE1; BND3; CD233; DI; EMPB3; EPB3; FR; MGC116750; MGC116753; MGC126619; MGC126623; RTA1A; SW; WD; WD1; WR | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title