missing translation for 'onlineSavingsMsg'
Learn More

Bambuterol Hydrochloride, TRC

Product Code. 30133846
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133846 100 mg 100mg
17931425 25 mg 25mg
17911075 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133846 Supplier Toronto Research Chemicals Supplier No. TRCB117500100MG
 Restricted Item
Estimated Shipment: 03-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81732-46-9
Molecular Formula C18 H29 N3 O5 . Cl H
Molecular Weight (g/mol) 403.9
Synonym Carbamic acid, dimethyl-, 5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-1,3-phenylene ester, monohydrochloride (9CI), Bambec, Bambuterol hydrochloride, KWD 2183
IUPAC Name [3-[2-(tert-butylamino)-1-hydroxyethyl]-5-(dimethylcarbamoyloxy)phenyl] N,N-dimethylcarbamate;hydrochloride
SMILES Cl.CN(C)C(=O)Oc1cc(OC(=O)N(C)C)cc(c1)C(O)CNC(C)(C)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H29N3O5.ClH/c1-18(2,3)19-11-15(22)12-8-13(25-16(23)20(4)5)10-14(9-12)26-17(24)21(6)7;/h8-10,15,19,22H,11H2,1-7H3;1H
Formula Weight 403.1874
Chemical Name or Material Bambuterol hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.