Learn More
Description
Bamboo TwinFlex™ D xg: Sustainability, comfort and sensitivity combined with high cut protection (CutD)
The patented Bamboo TwinFlex™ technology combines the best of both worlds in one fine 18-gauge cut protection glove (Cut Level D). On the inside of the glove, wearers experience noticeable comfort for their skin thanks to the bamboo-viscose. The smooth fit and high moisture absorption with a rapid action provides long-lasting comfort and high wearer acceptance. With a flexible fit thanks to the bamboo-viscose, the glove adapts precisely to fit the shape of the hand within 3 to 5 minutes of wearing. High-performance fibers offer a high level of protection against abrasion and cut hazards. The additional protection zone in the area between the thumb and index finger provides double the abrasion resistance (200%) compared to the inside of the glove to increase its service life/durability. Thanks to the Xtra-Grip aqua-polymer foam coating, the Bamboo TwinFlex™ D xg offers excellent grip in dry and slightly damp/oily conditions. In addition, the Bamboo TwinFlex D xg features a total of 45% sustainable materials: recycled polyamide and bamboo-viscose, which is based on renewable raw materials.
General Features:
- Excellent sensitivity in cut protection level D
- High level of wearer comfort thanks to bamboo fibre
- 45% sustainable materials: recycled polyamide and bamboo viscose, which is based on renewable raw materials
- High efficiency due to long service life, all thanks to protection + premium Xtra-Grip coating
- CO2-neutral production in Germany, free from solvents
Protective Features
- Certified to EN 388:2016 (4 X 4 1 D) and to EN 407 (X 1 X X X X)
- Dermatologically tested for skin compatibility, free from allergenic accelerators
- Particularly light, thin, and flexible with high cut protection (4X41D according to EN 388:2016)
- No need to remove the glove thanks to touchscreen capability protection zone with smooth material surface ensures trouble-free workflow in accordance with EN 407, the model is suitable for contact warmth up to +100 °C
Comfort Features
- Patented Bamboo TwinFlex™ yarn technology
- Silky soft feel against the skin
- Fast absorption and high retention of moisture
- Adaptive fit: Adapts precisely to fit the shape of any hand within 3 to 5 minutes of wearing
Areas of application
- Installation of sharp-edged or unburred parts
- Glass and metalworking industry, automotive industry
- Precision work
- Quality control
- Areas with use of touchscreens food processing
Specifications
Specifications
| Certifications/Compliance (Europe) | EN 388:2016 (4 X 4 1 D), EN 407 (X 1 X X X X) |
| Construction | Seamless Knit |
| Coating Material | Aqua-Polymer |
| Color | Green |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Bamboo-viscose, Elastane, HPPE, Polyamide, Steel |
| Product Type | Cut-resistant Gloves |
| Size | 8 |
| EN388 Standard | EN 388:4 X 4 1 D |
| Show More |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
