missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BAMBI Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148590
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
BAMBI (BMP and activin membrane-bound inhibitor homolog, NMA) is a membrane spanning glycoprotein that acts as a negative regulator of TGF-beta signaling during development. BAMBI transcription regulation is under the influence of beta-catenin, BMP, smad3 and smad4. BAMBI expression can increase in colorectal and hepatocellular carcinomas relative to non-cancerous tissues. BAMBI is expressed at high levels during odontogenesis. BAMBI is coexpressed with Bmp-4 during early Xenopus embryogenesis and can be detected in poorly metastatic human melanoma cell lines.
Specifications
| BAMBI | |
| Polyclonal | |
| Unconjugated | |
| BAMBI | |
| 2610003H06Rik; BAMBI; BMP and activin membrane bound inhibitor; BMP and activin membrane-bound inhibitor; BMP and activin membrane-bound inhibitor homolog; BMP and activin membrane-bound inhibitor, homolog; kinase-deficient TGFbeta superfamily receptor subunit; NMA; Non-metastatic gene A protein; putative transmembrane protein NMA | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 68010 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9D0L6 | |
| BAMBI | |
| Mouse myeloma cell line NS0-derived recombinant mouse BAMBI/NMA, Ile28-Thr143. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction