missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAMBI/NMA Rabbit anti-Human, Clone: 211, Novus Biologicals™
Rabbit Monoclonal Antibody
£307.00 - £448.00
Specifica
| Antigen | BAMBI/NMA |
|---|---|
| Clone | 211 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|
18254726
|
Novus Biologicals
NBP2-89698-50UL |
50 μg |
£307.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18237727
|
Novus Biologicals
NBP2-89698-100UL |
100 μg |
£448.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
BAMBI/NMA Monoclonal specifically detects BAMBI/NMA in Human samples. It is validated for ELISA.Specifica
| BAMBI/NMA | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 25805 | |
| IgG | |
| Protein A purified |
| 211 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| BMP and activin membrane-bound inhibitor homolog, BMP and activin membrane-bound inhibitor homolog (Xenopus laevis), NMANon-metastatic gene A protein, Putative transmembrane protein NMA | |
| Recombinant Human BAMBI/NMA Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title