missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ BAMBI/NMA Antibody Pair [HRP]

Product Code. p-7183261 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18240077

Brand: Novus Biologicals™ NBP27931715PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human BAMBI/NMA.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 9.375 pg/mL
Assay Sensitivity 9.38-600 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human BAMBI/NMA.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias BMP and activin membrane-bound inhibitor homolog, BMP and activin membrane-bound inhibitor homolog (Xenopus laevis), NMANon-metastatic gene A protein, Putative transmembrane protein NMA
Gene ID (Entrez) 25805
Gene Symbol BAMBI
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.5 mg/mL mouse anti-BAMBI/NMA monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.1.0 mg/mL of mouse anti-BAMBI/NMA monoclonal antibody,
  • Standard: Each vial contains 50 ng of recombinant BAMBI/NMA.
  • Quantity 15 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target BAMBI/NMA
    Technique 1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 600 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.