missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAMBI/NMA Antibody (OTI5D5), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71778AF750
This item is not returnable.
View return policy
Description
BAMBI/NMA Monoclonal specifically detects BAMBI/NMA in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| BAMBI/NMA | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| BAMBI | |
| Recombinant protein fragment corresponding to amino acids 21-152 of human BAMBI/NMA (NP_036474) produced in E.coli. | |
| 0.1 mL | |
| 25805.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI5D5 | |
| Western Blot | |
| BMP and activin membrane-bound inhibitor homolog, BMP and activin membrane-bound inhibitor homolog (Xenopus laevis), NMANon-metastatic gene A protein, Putative transmembrane protein NMA | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction