missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ BAMBI/NMA Antibody (3C1-1D1), Novus Biologicals™

Product Code. 18347969 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347969 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347969 Supplier Novus Biologicals™ Supplier No. H00025805M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BAMBI/NMA Monoclonal antibody specifically detects BAMBI/NMA in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Spécification

Antigen BAMBI/NMA
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 3C1-1D1
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH19252
Gene Alias BMP and activin membrane-bound inhibitor homolog, BMP and activin membrane-bound inhibitor homolog (Xenopus laevis), NMANon-metastatic gene A protein, Putative transmembrane protein NMA
Host Species Mouse
Immunogen BAMBI (AAH19252, 1 a.a. ∽ 260 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MDRHSSYIFIWLQLELCAMAVLLTKGEIRCYCDAAHCVATGYMCKSELSACFSRLLDPQNSNSPLTHGCLDSLASTTDICQAKQARNHSGTTIPTLECCHEDMCNYRGLHDVLSPPRGEASGQGNRYQHDGSRNLITKVQELTSSKELWFRAAVIAVPIAGGLTLVLLIMLALRMLRSENKRLQDQRQQMLSRLHYSFHGHHSKKGQVAKLDLECMVPVSGHENCCLTCDKMRQADLSNDKILSLVHWGMYSGHGKLEFV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25805
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.