missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAMBI/NMA Antibody (211), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89698PE
This item is not returnable.
View return policy
Description
BAMBI/NMA Monoclonal antibody specifically detects BAMBI/NMA in Human samples. It is validated for ELISA
Specifications
| BAMBI/NMA | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 25805 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| 211 | |
| ELISA | |
| BMP and activin membrane-bound inhibitor homolog, BMP and activin membrane-bound inhibitor homolog (Xenopus laevis), NMANon-metastatic gene A protein, Putative transmembrane protein NMA | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human BAMBI/NMA (Accession#: NP_036474.1; Met 1-Ala 152). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction