missing translation for 'onlineSavingsMsg'
Learn More

Balzers chrome layer etching solution, Fluka™

Product Code. p-7230404 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 L
200 L
Packaging:
Metal Drum
Plastic Pail
Unit Size:
200 L
25 L
This item is not returnable. View return policy

Product Code. 15677030

Brand: Honeywell-Fluka 7472725L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Balzers chrome layer etching solution

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Balzers chrome layer etching solution
Color Light Yellow
CAS 16774-21-3, 64-19-7, 7732-18-5
Boiling Point 100°C (1.013 hPa)
Grade Synthesis
Packaging Plastic Pail
pH Acidic
Quantity 25 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.