missing translation for 'onlineSavingsMsg'
Learn More

Balsalazide, MedChemExpress

Product Code. 16488568
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488568 10 mg 10mg
16408416 100 mg 100mg
16404447 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488568 Supplier Medchem Express Supplier No. HYB0667
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Balsalazide could suppress colitis-associated carcinogenesis through modulation of IL-6/STAT3 pathway.

Balsalazide could suppress colitis-associated carcinogenesis through modulation of IL-6/STAT3 pathway.

Specifications

Chemical Name or Material Balsalazide
CAS 80573-04-2
Color Orange
Purity Grade Notes Research
Molecular Formula C17H15N3O6
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (279.86 mM; Need ultrasonic)
SMILES O=C(O)C1=CC(/N=N/C2=CC=C(C(NCCC(O)=O)=O)C=C2)=CC=C1O
Molecular Weight (g/mol) 357.32
Formula Weight 357.32
Percent Purity 99.2%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.