missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Baloxavir Marboxil, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Baloxavir Marboxil |
| CAS | 1985606-14-1 |
| Molecular Formula | C27 H23 F2 N3 O7 S |
| Quantity | 250 mg |
| Synonym | Baloxavir Marboxil, Carbonic acid, [[(12aR)-12-[(11S)-7,8-difluoro-6,11-dihydrodibenzo[b,e]thiepin-11-yl]-3,4,6,8,12,12a-hexahydro-6,8-dioxo-1H-[1,4]oxazino[3,4-c]pyrido[2,1-f][1,2,4]triazin-7-yl]oxy]methyl methyl ester |
| InChI Formula | InChI=1S/C27H23F2N3O7S/c1-36-27(35)39-14-38-25-19(33)8-9-31-24(25)26(34)30-10-11-37-12-21(30)32(31)23-15-6-7-18(28)22(29)17(15)13-40-20-5-3-2-4-16(20)23/h2-9,21,23H,10-14H2,1H3/t21-,23+/m1/s1 |
| SMILES | COC(=O)OCOC1=C2N(C=CC1=O)N([C@@H]3COCCN3C2=O)[C@H]4c5ccc(F)c(F)c5CSc6ccccc46 |
| IUPAC Name | (((R)-12-((S)-7,8-difluoro-6,11-dihydrodibenzo[b,e]thiepin-11-yl)-6,8-dioxo-3,4,6,8,12,12a-hexahydro-1H-[1,4]oxazino[3,4-c]pyrido[2,1-f][1,2,4]triazin-7-yl)oxy)methyl methyl carbonate |
| Molecular Weight (g/mol) | 571.549 |
| Formula Weight | 571.122 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction