Learn More
Lenz Laborglasintrumente Ball Flanges KF, Socket with Olive
Click to view available options
Standard Taper Joint:
Socket KF 15
Unit Size:
Each
Description
- With KF socket
- With glass olive
- Made of DURAN™ tubing
Specifications
Specifications
| Includes | Olive, 16 mm |
| Standard Taper Joint | Socket KF 15 |