missing translation for 'onlineSavingsMsg'
Learn More

Balipodect, MedChemExpress

Product Code. 16485188
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485188 10 mg 10mg
16481407 50 mg 50mg
16437917 5 mg 5mg
16425886 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485188 Supplier Medchem Express Supplier No. HY12472

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Balipodect (TAK-063) is a highly potent, selective and orally active PDE10A inhibitor with IC50 of 0.30 nM; >15000-fold selectivity over other PDEs.

Balipodect (TAK-063) is a highly potent, selective and orally active PDE10A inhibitor with IC50 of 0.30 nM

Specifications

Chemical Name or Material Balipodect
CAS 1238697-26-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H17FN6O2
Quantity 10 mg
Synonym TAK-063
Solubility Information DMSO : 25 mg/mL (58.35 mM; Need ultrasonic)
SMILES FC1=CC(N2N=CC=C2)=CC=C1N3N=C(C4=CC=NN4C5=CC=CC=C5)C(C(OC)=C3)=O
Molecular Weight (g/mol) 428.42
Formula Weight 428.42
Percent Purity 98.69%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.