Learn More
Mettler Toledo Balance Classic Light PL6000-S/13 top pan capacity6100g 240V 50-60Hz weighing pan diameter 160mm
Brand:  Mettler Toledo 12110038
Balance Classic Light PL6000-S/13 top pan capacity6100g 240V 50-60Hz weighing pan diameter 160mm
Product Code. 12784189