Learn More
Avantor™ BAKERBOND Speedisk™ SAX Quaternary Amine Extraction Disk
Maximize capacity, adsorption and SPE speed. Avantor BAKERBOND Speedisk SAX Quaternary Amine Extraction Disk are neither cartridge nor membrane that provides an excellent SPE tool for environmental samples and combines a thin bed of BAKERBOND sorbent with a special laminar configuration for testing drinking water, ground water, waste water, solid waste or process streams.
Brand: Avantor™ 8058-06
Code : Z2
Additional Details : Weight : 1.00000kg
Description
- Pre-assembled with rigid support and baffle to maximize laminar flow, allow for filtration, resist clogging, and maximize capacity, adsorption, and speed
- Require minimal glassware, set up time, and assembly
- Minimizes impact of extractable compounds
- Resists clogging and ensures high throughput rate
- Reduce SPE run times
Specifications
EPA Method 552.1, Haloacetic Acids and Dalapon | |
SAX Quaternary Amine Extraction Disk | |
50 mm |
Expanded Extraction Station (8095-06) | |
20/Pk. |