missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5109442
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form oligomers or heterodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein localizes to mitochondria, and functions to induce apoptosis. It interacts with and accelerates the opening of the mitochondrial voltage-dependent anion channel, which leads to a loss in membrane potential and the release of cytochrome c. This protein also interacts with the tumor suppressor P53 after exposure to cell stress.
Specifications
| BAK1 | |
| Polyclonal | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08734, Q16611 | |
| BAK1 | |
| A synthetic peptide of human BAK1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| BAK1 | |
| Apoptosis regulator BAK; BAK; BAK1; BAK-LIKE; Bcl-2 homologous antagonist/killer; BCL2L7; Bcl2-L-7; BCL2-like 7protein; Bcl-2-like protein 7; CDN1; MGC117255; MGC3887; N-Bak; N-BAK1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116502, 12018, 578 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction