missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77152SS
This item is not returnable.
View return policy
Description
BAK Polyclonal specifically detects BAK in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| BAK | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Apoptosis regulator BAK, BAK-LIKE, BAKMGC3887, BCL2-antagonist/killer 1, Bcl2-L-7, BCL2L7bcl-2 homologous antagonist/killer, BCL2-like 7 protein, Bcl-2-like protein 7, CDN1, MGC117255, pro-apoptotic protein BAK | |
| Rabbit | |
| Ion exchange chromatography | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 578 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1-2 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2 ug/ml | |
| Q16611 | |
| BAK1 | |
| Antibody was raised against a peptide corresponding to 13 amino acids near the amino-terminus of human Bak. The immunogen is located within amino acids 30 - 80 of Bak. Amino Acid Squence: QEQEAEGVAAPAD | |
| 0.025 mg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction