missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAK Antibody (38E2), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-74026IR
This item is not returnable.
View return policy
Description
BAK Monoclonal specifically detects BAK in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| BAK | |
| Monoclonal | |
| DyLight 755 | |
| NP_001179 | |
| BAK1 | |
| Recombinant human BAK1 (29-187aa) purified from E.coli | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 578 | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 8B4 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| Apoptosis regulator BAK, BAK-LIKE, BAKMGC3887, BCL2-antagonist/killer 1, Bcl2-L-7, BCL2L7bcl-2 homologous antagonist/killer, BCL2-like 7 protein, Bcl-2-like protein 7, CDN1, MGC117255, pro-apoptotic protein BAK | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human BAK1 mAb, clone 8B4, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human BAK1 protein. | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction