missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAIAP3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£141.00 - £398.00
Specifications
| Antigen | BAIAP3 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18622016
|
Novus Biologicals
NBP2-41079-0.025mg |
0.025 mg |
£141.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18151189
|
Novus Biologicals
NBP2-41079 |
0.1 mg |
£398.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAIAP3 Polyclonal specifically detects BAIAP3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| BAIAP3 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 8938 | |
| Antibody was raised against a 12 amino acid synthetic peptide from near the amino terminus of human BAP3. The immunogen is located within the first 50 amino acids of BAP3. Amino Acid Squence: GAIWRSRSPAMS | |
| Primary |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| BAI1-associated protein 3, BAI-associated protein 3, BAP3KIAA0734MGC138334 | |
| BAIAP3 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title