missing translation for 'onlineSavingsMsg'
Learn More

BAIAP2L1 Antibody (4A7), Novus Biologicals™

Product Code. 18394989 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394989 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394989 Supplier Novus Biologicals Supplier No. H00055971M09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BAIAP2L1 Monoclonal antibody specifically detects BAIAP2L1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAIAP2L1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4A7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_061330.2
Gene Alias BAI1-associated protein 2-like 1, BAI1-associated protein 2-like protein 1, brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 1, FLJ42275, IRTKSInsulin receptor tyrosine kinase substrate
Host Species Mouse
Immunogen BAIAP2L1 (NP_061330.2, 1 a.a. ~ 511 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSRGPEEVNRLTESTYRNVMEQFNPGLRNLINLGKNYEKAVNAMILAGKAYYDGVAKIGEIATGSPVSTELGHVLIEISSTHKKLNESLDENFKKFHKEIIHELEKKIELDVKYMNATLKRYQTEHKNKLESLEKSQAELKKIRRKSQGSRNALKYEHKEIEYVETVTSRQSEIQKFIADGCKEALLEEKRRFCFLVDKHCGFANHIHYYHLQSAELLNSKLPRWQETCVDAIKVPEKIMNMIEEIKTPASTPVSGTPQASPMIERSNVVRKDYDTLSKCSPKMPPAPSGRAYTSPLIDMFNNPATAAPNSQRVNNSTGTSEDPSLQRSVSVATGLNMMKKQKVKTIFPHTAGSNKTLLSFAQGDVITLLIPEEKDGWLYGEHDVSKARGWFPSSYTKLLEENETEAVTVPTPSPTPVRSISTVNLSENSSVVIPPPDYLECLSMGAAADRRADSARTTSTFKAPASKPETAAPNDANGTAKPPFLSGENPFATVKLRPTVTNDRSAPIIR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55971
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.