missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ BAIAP2 HEK293 Cell Transient Overexpression Lysate (Nondenatured)

Product Code. 16160904
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 16160904

Brand: Abnova™ L042T6.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Human overexpression lysate

Specifications

Cell Line HEK293
For Use With (Application) Immunoprecipitation, Western Blot
Description BAIAP2 HEK293 Cell Transient Overexpression Lysate(Non-Denatured)
Gene Accession No. BC032559
Lysis Buffer Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF.
Molecular Weight (g/mol) Theoretical M.W. 57kDa
Plasmid pCMV-BAIAP2 full length
Quality Control Testing Transient overexpression cell lysate was tested with anti-BAIAP2 antibody (Mfr. No. H00010458-M01) by Western Blot.
Quantity 100 μg
Storage Buffer In modified RIPA Lysis Buffer.
Host Species Human
Concentration 2 mg/mL
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.