missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAIAP2 Antibody (OTI 1D9), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71603PE
This item is not returnable.
View return policy
Description
BAIAP2 Monoclonal specifically detects BAIAP2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| BAIAP2 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| BAIAP2 | |
| Full length recombinant protein of human BAIAP2 (NP_006331) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI 1D9 | |
| Flow Cytometry | |
| BAI1-associated protein 2IRS-58, BAI-associated protein 2, BAP2, brain-specific angiogenesis inhibitor 1-associated protein 2, Fas ligand-associated factor 3, FLAF3, Insulin receptor substrate p53, Insulin receptor substrate p53/p58, Insulin receptor substrate protein of 53 kDa, IRSp53, IRSp53/58, Protein BAP2 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 10458.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction