missing translation for 'onlineSavingsMsg'
Learn More

BAIAP2 Antibody (3F3-2D3), Novus Biologicals™

Product Code. 18367508 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18367508 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18367508 Supplier Novus Biologicals Supplier No. H00010458M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BAIAP2 Monoclonal antibody specifically detects BAIAP2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAIAP2
Applications Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA
Classification Monoclonal
Clone 3F3-2D3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH32559
Gene Alias BAI1-associated protein 2IRS-58, BAI-associated protein 2, BAP2, brain-specific angiogenesis inhibitor 1-associated protein 2, Fas ligand-associated factor 3, FLAF3, Insulin receptor substrate p53, Insulin receptor substrate p53/p58, Insulin receptor substrate protein of 53 kDa, IRSp53, IRSp53/58, Protein BAP2
Host Species Mouse
Immunogen BAIAP2 (AAH32559, 1 a.a. ~ 512 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSLSRSEEMHRLTENVYKTIMEQFNPSLRNFIAMGKNYEKALAGVTYAAKGYFDALVKMGELASESQGSKELGDVLFQMAEVHRQIQNQLEEMLKSFHNELLTQLEQKVELDSRYLSAALKKYQTEQRSKGDALDKCQAELKKLRKKSQGSKNPQKYSDKELQYIDAISNKQGELENYVSDGYKTALTEERRRFCFLVEKQCAVAKNSAAYHSKGKELLAQKLPLWQQACADPSKIPERAVQLMQQVASNGATLPSALSASKSNLVISDPIPGAKPLPVPPELAPFVGRMSAQESTPIMNGVTGPDGEDYSPWADRKAAQPKSLSPPQSQSKLSDSYSNTLPVRKSVTPKNSYATTENKTLPRSSSMAAGLERNGRMRVKAIFSHAAGDNSTLLSFKEGDLITLLVPEARDGWHYGESEKTKMRGWFPFSYTRVLDSDGSDRLHMSLQQGKSSSTGNLLDKDDLAIPPPDYGAASRAFPAQTASGFKQRPYSVAVPAFSQGLDDYGARSMSR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10458
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.