missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BAI3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567719
This item is not returnable.
View return policy
Description
Predicted to react with Mouse samples.
This p53-target gene encodes a brain-specific angiogenesis inhibitor, a seven-span transmembrane protein, and is thought to be a member of the secretin receptor family. Brain-specific angiogenesis proteins BAI2 and BAI3 are similar to BAI1 in structure, have similar tissue specificities, and may also play a role in angiogenesis.
Specifications
| BAI3 | |
| Polyclonal | |
| Unconjugated | |
| Adgrb3 | |
| A830096D10Rik; ADGRB3; Adhesion G protein-coupled receptor B3; BAI3; Brain-specific angiogenesis inhibitor 3; dJ91B17.1 (brain-specific angiogenesis inhibitor 3); G protein-coupled receptor BAI3; KIAA0550 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 210933, 577 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 30% glycerol, 0.87% NaCl and 0.01% sodium azide; pH 7.3 | |
| O60242, Q80ZF8 | |
| Adgrb3 | |
| Synthesized peptide derived from human BAI3. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction